Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303.3 | NP_001294.2 | 3 | 684 | ||
GenBank mRNA | AK295925.1 | - | 3 | 482 | ||
DA200545.1 | - | 1 | 31 |