Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPSE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166244.1 | NP_001159716.1 | 8 | 1146 | ||
NM_001166245.1 | NP_001159717.1 | 7 | 984 | |||
NM_001166246.1 | NP_001159718.1 | 9 | 1320 | |||
NM_021828.4 | NP_068600.4 | 9 | 1320 | |||
XM_006717937.2 | XP_006718000.1 | 9 | 1380 | |||
XM_011540029.1 | XP_011538331.1 | 9 | 1320 | |||
XM_011540030.1 | XP_011538332.1 | 8 | 1145 | |||
XM_011540031.2 | XP_011538333.1 | 8 | 929 | |||
XM_011540033.2 | XP_011538335.1 | 8 | 953 | |||
XM_017016495.1 | XP_016871984.1 | 9 | 1320 | |||
XM_017016497.1 | XP_016871986.1 | 8 | 913 | |||
XM_017016498.1 | XP_016871987.1 | 8 | 1214 | |||
GenBank mRNA | AF282885.1 | - | 7 | 971 | ||
AF282886.1 | - | 8 | 1133 | |||
AF282887.1 | - | 9 | 1307 | |||
AJ299719.1 | - | 9 | 1307 | |||
AJ299720.1 | - | 9 | 1247 | |||
AK094037.1 | - | 8 | 1078 | |||
AK314759.1 | - | 9 | 1320 | |||
BC112356.1 | - | 9 | 1289 | 40033830 |