Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASDHPPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015423.2 | NP_056238.2 | 4 | 743 | ||
GenBank mRNA | AF136978.1 | - | 4 | 765 | ||
AF151057.1 | - | 4 | 763 | |||
AF151838.1 | - | 4 | 763 | |||
AF201943.1 | - | 4 | 712 | |||
AF302110.1 | - | 4 | 763 | |||
AK293362.1 | - | 3 | 504 | |||
AK312529.1 | - | 4 | 647 | |||
AL050073.1 | - | 3 | 304 | |||
BC015470.1 | - | 4 | 634 | 3914010 | ||
BC016728.1 | - | 4 | 663 | 4081713 |