Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320975.1 | NP_001307904.1 | 8 | 2811 | ||
NM_001320976.1 | NP_001307905.1 | 9 | 2883 | |||
NM_004376.6 | NP_004367.2 | 8, 9 | 1727 | |||
NM_078470.5 | NP_510870.1 | 9 | 2966 | |||
XM_011539298.2 | XP_011537600.1 | 7, 8 | 1037 | |||
GenBank mRNA | BC078161.1 | - | 9 | 2378 | 6150426 |