Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001153446.1 | NP_001146918.1 | 3 | 154 | ||
NM_001153484.1 | NP_001146956.1 | 2 | 247 | |||
NM_001153552.1 | NP_001147024.1 | 2 | 247 | |||
NM_001153585.1 | NP_001147057.1 | 2 | 287 | |||
NM_001153635.1 | NP_001147107.1 | 3 | 154 | |||
NM_001153663.1 | NP_001147135.1 | 2 | 247 | |||
NM_001153690.1 | NP_001147162.1 | 2 | 287 | |||
NM_032547.2 | NP_115936.2 | 3 | 252 | |||
GenBank mRNA | AB015335.1 | - | 2 | 118 | ||
AF330205.1 | - | 2 | 138 | |||
AF448857.1 | - | 2 | 138 | |||
AK027797.1 | - | 3 | 252 | |||
BC016511.1 | - | 2 | 89 | |||
BC062684.1 | - | 3 | 154 | 4691440 | ||
BU070009.1 | - | 2 | 226 | |||
CN367654.1 | - | 2 | 247 | |||
DA625782.1 | - | 2 | 287 | |||
DA695503.1 | - | 2 | 221 | |||
DB204055.1 | - | 3 | 154 | |||
DB465370.1 | - | 2 | 196 |