Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001153446.1 | NP_001146918.1 | 2 | 100 | ||
NM_001153585.1 | NP_001147057.1 | 1 | 233 | |||
NM_001153635.1 | NP_001147107.1 | 2 | 100 | |||
NM_001153690.1 | NP_001147162.1 | 1 | 233 | |||
NM_032547.2 | NP_115936.2 | 2 | 198 | |||
GenBank mRNA | AK027797.1 | - | 2 | 198 | ||
BC062684.1 | - | 2 | 100 | 4691440 | ||
BU070009.1 | - | 1 | 172 | |||
DA625782.1 | - | 1 | 233 | |||
DB204055.1 | - | 2 | 100 |