Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172684.1 | NP_001166155.1 | 8 | 1072 | ||
NM_001172685.1 | NP_001166156.1 | 7 | 827 | |||
NM_052840.4 | NP_443072.3 | 8 | 1310 | |||
GenBank mRNA | AF401233.1 | - | 8 | 1255 | ||
AF425606.1 | - | 8 | 1071 | |||
AK124858.1 | - | 7 | 864 | |||
AK131098.1 | - | 10 | 2459 | |||
AK294382.1 | - | 7 | 827 | |||
AK296004.1 | - | 8 | 1008 | |||
BC030835.1 | - | 8 | 1013 | 4793742 | ||
BC033838.2 | - | 8 | 914 | 4214670 |