Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172684.1 | NP_001166155.1 | 10 | 1183 | ||
NM_001172685.1 | NP_001166156.1 | 9 | 956 | |||
NM_052840.4 | NP_443072.3 | 11 | 1502 | |||
GenBank mRNA | AF401233.1 | - | 11 | 1447 | ||
AF425606.1 | - | 11 | 1263 | |||
AK124858.1 | - | 10 | 1056 | |||
AK131098.1 | - | 13 | 2651 | |||
AK294382.1 | - | 9 | 956 | |||
AK296004.1 | - | 10 | 1119 | |||
BC030835.1 | - | 11 | 1205 | 4793742 | ||
BC033838.2 | - | 11 | 1106 | 4214670 |