Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021942.5 | NP_068761.4 | 10 | 1219 | ||
NM_199053.2 | NP_951008.1 | 10 | 1219 | |||
XM_017008537.1 | XP_016864026.1 | 10 | 1219 | |||
XM_017008538.1 | XP_016864027.1 | 10 | 1219 | |||
XM_017008539.1 | XP_016864028.1 | 9 | 964 | |||
GenBank mRNA | AK091146.1 | - | 5 | 443 | ||
AK096345.1 | - | 10 | 1193 | |||
AK315057.1 | - | 5 | 425 | |||
AL136752.1 | - | 2 | 128 | |||
AL833451.1 | - | 9 | 1027 | |||
AL833571.1 | - | 10 | 1198 | |||
BC051724.1 | - | 10 | 1163 | 5265781 | ||
BX538044.1 | - | 10 | 1173 | |||
BX647127.1 | - | 10 | 1190 | |||
BX648499.1 | - | 10 | 1194 | |||
CR533553.1 | - | 2 | 63 |