Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000096.3 | NP_000087.1 | 9 | 1841 | ||
NR_046371.1 | 9 | 1841 | ||||
XM_006713499.2 | XP_006713562.1 | 9 | 1846 | |||
XM_006713500.3 | XP_006713563.1 | 9 | 1846 | |||
XM_006713501.2 | XP_006713564.1 | 9 | 1846 | |||
XM_011512435.1 | XP_011510737.1 | 9 | 1846 | |||
XM_017005734.1 | XP_016861223.1 | 9 | 1846 | |||
XM_017005735.1 | XP_016861224.1 | 9 | 1846 | |||
GenBank mRNA | AK095290.1 | - | 8 | 1130 | ||
AK291219.1 | - | 9 | 1623 | |||
AK299272.1 | - | 9 | 1627 | |||
AK309279.1 | - | 5 | 813 | |||
BC094709.1 | - | 9 | 1625 | |||
BC142714.1 | - | 9 | 1597 | 40148840 | ||
BC146663.1 | - | 9 | 1597 | 40148048 | ||
BC146801.1 | - | 9 | 1621 | |||
M13536.1 | - | 6 | 938 | |||
M13699.1 | - | 9 | 1588 |