Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021943.2 | NP_068762.1 | 5 | 798 | ||
XM_005249269.1 | XP_005249326.1 | 4 | 767 | |||
XM_017011171.1 | XP_016866660.1 | 4 | 771 | |||
GenBank mRNA | AK023284.1 | - | 5 | 472 | ||
BC031481.2 | - | 5 | 414 | 4448968 | ||
BP305887.1 | - | 5 | 527 |