Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCORL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184772.2 | NP_001171701.1 | 3 | 2305 | ||
NM_021946.4 | NP_068765.3 | 3 | 2305 | |||
XM_005262452.4 | XP_005262509.2 | 5 | 2721 | |||
XM_005262453.4 | XP_005262510.2 | 5 | 2489 | |||
XM_005262454.3 | XP_005262511.2 | 4 | 2433 | |||
XM_005262455.4 | XP_005262512.2 | 4 | 2376 | |||
XM_005262456.4 | XP_005262513.2 | 5 | 2721 | |||
XM_006724776.3 | XP_006724839.1 | 5 | 2506 | |||
XM_006724777.3 | XP_006724840.1 | 4 | 2401 | |||
XM_006724779.2 | XP_006724842.1 | 5 | 2592 | |||
XM_017029721.1 | XP_016885210.1 | 5 | 2752 | |||
XM_017029722.1 | XP_016885211.1 | 4 | 2806 | |||
GenBank mRNA | AK074089.1 | - | 3 | 2218 | ||
AL096777.1 | - | 1 | 567 |