Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004075.4 | NP_004066.1 | 9 | 2341 | ||
XM_017018832.1 | XP_016874321.1 | 10 | 2476 | |||
GenBank mRNA | AK098615.1 | - | 4 | 745 | ||
AK125915.1 | - | 11 | 2004 | |||
AK290552.1 | - | 9 | 1641 | |||
BC017466.1 | - | 9 | 1764 | |||
BC030519.1 | - | 9 | 2014 | 5169081 | ||
D83702.1 | - | 9 | 2031 | |||
D84657.1 | - | 9 | 1445 |