Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001868.3 | NP_001859.1 | 3 | 496 | ||
GenBank mRNA | AK131054.1 | - | 831 | |||
AK291493.1 | - | 3 | 373 | |||
AK293366.1 | - | 3 | 308 | |||
BC005279.1 | - | 3 | 368 | 3949850 | ||
BT007313.1 | - | 3 | 346 | |||
DC378103.1 | - | 3 | 496 | |||
X67318.1 | - | 3 | 353 |