Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001868.3 | NP_001859.1 | 9 | 1213 | ||
GenBank mRNA | AK124421.1 | - | 2 | 1641 | ||
AK131054.1 | - | 591 | ||||
AK291493.1 | - | 9 | 1090 | |||
BC005279.1 | - | 9 | 1085 | 3949850 | ||
BT007313.1 | - | 9 | 1063 | |||
X67318.1 | - | 9 | 1070 |