Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM204A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134672.1 | NP_001128144.1 | 2 | 199 | ||
NM_022063.2 | NP_071346.1 | 3 | 400 | |||
XM_005270024.1 | XP_005270081.1 | 4 | 577 | |||
GenBank mRNA | AK023250.1 | - | 3 | 379 | ||
AK026630.1 | - | 3 | 393 | |||
BC023577.2 | - | 3 | 383 | 4765095 | ||
BM837313.1 | - | 2 | 192 | |||
CR457304.1 | - | 1 | 133 | |||
DA168924.1 | - | 3 | 404 |