Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022064.4 | NP_071347.2 | 17, 18 | 1639 | ||
NR_135218.1 | 17, 18 | 1639 | ||||
XM_006713285.1 | XP_006713348.1 | 17, 18 | 1797 | |||
XM_011533995.1 | XP_011532297.1 | 17, 18 | 1574 | |||
XM_017007018.1 | XP_016862507.1 | 17, 18 | 1601 | |||
GenBank mRNA | AF370367.1 | - | 8, 9 | 793 | ||
AK090632.1 | - | 7, 8 | 681 | |||
AK096353.1 | - | 17, 18 | 1553 | |||
AK308130.1 | - | 17, 18 | 1581 | |||
AK308607.1 | - | 14, 15 | 1317 | |||
AL832034.1 | - | 17, 18 | 1601 | |||
AL832138.1 | - | 17, 18 | 1576 | |||
AL832344.1 | - | 17, 18 | 1578 | |||
AY744152.1 | - | 16, 17 | 1522 | |||
BC041145.1 | - | 2, 3 | 172 | |||
BC057392.1 | - | 4, 5 | 329 | |||
BC130632.1 | - | 17, 18 | 1580 | 40146663 | ||
CR627051.1 | - | 15, 16 | 2147 |