Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022064.4 | NP_071347.2 | 39 | 4033 | ||
NR_135218.1 | 39 | 4273 | ||||
XM_006713285.1 | XP_006713348.1 | 39 | 4191 | |||
XM_011533995.1 | XP_011532297.1 | 39 | 3968 | |||
XM_017007018.1 | XP_016862507.1 | 39 | 3995 | |||
GenBank mRNA | AF370367.1 | - | 30 | 3187 | ||
AK022627.1 | - | 17 | 1859 | |||
AK090632.1 | - | 29 | 3075 | |||
AK096353.1 | - | 39 | 3947 | |||
AK126883.1 | - | 23 | 3740 | |||
AL136729.1 | - | 12 | 1417 | |||
AL832034.1 | - | 39 | 3995 | |||
AL832138.1 | - | 38 | 4284 | |||
AL832344.1 | - | 39 | 3971 | |||
AY744152.1 | - | 38 | 3916 | |||
BC041145.1 | - | 24 | 2566 | |||
BC057392.1 | - | 26 | 2723 | |||
BC088801.1 | - | 21 | 2294 | |||
BC130632.1 | - | 39 | 3974 | 40146663 | ||
CR627051.1 | - | 35 | 5605 |