Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022066.3 | NP_071349.3 | 6 | 829 | ||
XM_005257567.1 | XP_005257624.1 | 6 | 813 | |||
XM_017024934.1 | XP_016880423.1 | 6 | 813 | |||
XM_017024935.1 | XP_016880424.1 | 6 | 813 | |||
GenBank mRNA | AB051521.1 | - | 6 | 829 |