Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM111A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142519.2 | NP_001135991.1 | 5 | 1992 | ||
NM_001142520.2 | NP_001135992.1 | 5 | 2015 | |||
NM_001142521.2 | NP_001135993.1 | 5 | 1828 | |||
NM_001312909.1 | NP_001299838.1 | 6 | 2230 | |||
NM_001312910.1 | NP_001299839.1 | 5 | 2054 | |||
NM_001312911.1 | NP_001299840.1 | 4 | 1819 | |||
NM_022074.3 | NP_071357.2 | 4 | 1726 | |||
NM_198847.2 | NP_942144.1 | 4 | 1687 | |||
GenBank mRNA | AB067482.1 | - | 2 | 2130 | ||
AK025319.1 | - | 1 | 148 | |||
AK026447.1 | - | 1 | 337 | |||
AK092061.1 | - | 4 | 1670 | |||
AK092953.1 | - | 1 | 906 | |||
AK130799.1 | - | 1 | 47 | |||
AK291453.1 | - | 4 | 1692 | |||
BC013137.1 | - | 1 | 922 | |||
BC054515.1 | - | 1 | 231 | |||
BC071759.1 | - | 4 | 1686 | 6451741 | ||
CR749358.1 | - | 3 | 2333 |