Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022078.2 | NP_071361.2 | 3 | 1041 | ||
GenBank mRNA | AK022517.1 | - | 3 | 1036 | ||
AK057930.1 | - | 3 | 1036 | |||
AK303183.1 | - | 4 | 984 | |||
AK308100.1 | - | 3 | 904 | |||
AL833886.1 | - | 3 | 1023 | |||
BC007767.2 | - | 3 | 1017 | 4298740 | ||
CR936767.1 | - | 3 | 1041 |