Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199322.1 | NP_001186251.1 | 5, 6 | 533 | ||
NM_001199323.1 | NP_001186252.1 | 3, 4 | 300 | |||
NM_201280.2 | NP_958437.1 | 4, 5 | 430 | |||
GenBank mRNA | AF426434.1 | - | 4, 5 | 430 | ||
AK025442.1 | - | 4, 5 | 383 | |||
AK301142.1 | - | 4, 5 | 431 | |||
BC032438.1 | - | 4, 5 | 393 | |||
BC119644.1 | - | 4, 5 | 415 | 40115738 | ||
BC119645.1 | - | 5, 6 | 515 | 40115739 | ||
BM829259.1 | - | 3, 4 | 271 | |||
BX647336.1 | - | 5, 6 | 602 | |||
CR749569.1 | - | 4, 5 | 405 |
Gene Symbol: EEF1E1-BLOC1S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037618.1 | 6, 7 | 738 | |||
GenBank mRNA | BI552521.1 | - | 6, 7 | 678 |