Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199322.1 | NP_001186251.1 | 6 | 1067 | ||
NM_001199323.1 | NP_001186252.1 | 4 | 834 | |||
NM_201280.2 | NP_958437.1 | 5 | 964 | |||
GenBank mRNA | AF426434.1 | - | 5 | 964 | ||
AK025442.1 | - | 5 | 917 | |||
AK301142.1 | - | 5 | 965 | |||
AK311385.1 | - | 2 | 815 | |||
BC032438.1 | - | 5 | 927 | |||
BC119644.1 | - | 5 | 949 | 40115738 | ||
BC119645.1 | - | 6 | 1049 | 40115739 | ||
BX647336.1 | - | 6 | 1136 | |||
CR749569.1 | - | 5 | 939 |
Gene Symbol: EEF1E1-BLOC1S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037618.1 | 7 | 1272 |