Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318253.1 | NP_001305182.1 | 21 | 2650 | ||
NM_133171.4 | NP_573403.1 | 21 | 2582 | |||
NM_182764.2 | NP_877496.1 | 20 | 2478 | |||
XM_005260496.3 | XP_005260553.1 | 21 | 2147 | |||
XM_005260498.3 | XP_005260555.1 | 20 | 2509 | |||
XM_005260499.3 | XP_005260556.1 | 20 | 2480 | |||
XM_005260500.3 | XP_005260557.1 | 19 | 2008 | |||
XM_005260501.4 | XP_005260558.1 | 20 | 2018 | |||
XM_006723854.3 | XP_006723917.1 | 20 | 2081 | |||
XM_017028009.1 | XP_016883498.1 | 20 | 2453 | |||
XM_017028010.1 | XP_016883499.1 | 19 | 1956 | |||
XM_017028011.1 | XP_016883500.1 | 19 | 2409 | |||
GenBank mRNA | AB058737.1 | - | 20 | 2093 | ||
AF398886.1 | - | 19 | 2021 | |||
AF417861.1 | - | 19 | 1953 | |||
AK021718.1 | - | 10 | 1149 | |||
AK022731.1 | - | 8 | 864 | |||
AK023103.1 | - | 9 | 909 | |||
AK092796.1 | - | 1 | 658 | |||
AK293861.1 | - | 19 | 2046 | |||
AK299318.1 | - | 23 | 2193 | |||
AK302710.1 | - | 11 | 1385 | |||
AL713765.1 | - | 9 | 2595 | |||
BC000143.4 | - | 20 | 2158 | 3047992 | ||
BC021079.2 | - | 10 | 1145 | |||
BX648819.1 | - | 20 | 2059 |