Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBED8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303251.1 | NP_001290180.1 | 3 | 2319 | ||
NM_022090.4 | NP_071373.2 | 2 | 2245 | |||
GenBank mRNA | AK026754.1 | - | 1 | 2051 | ||
BC032603.1 | - | 3 | 2266 | 5535434 | ||
BC112128.1 | - | 1 | 1787 | 8327596 | ||
BC112130.1 | - | 1 | 1787 | 8327598 | ||
BM903536.1 | - | 1 | 264 |