Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022095.3 | NP_071378.1 | 8, 9 | 1482 | ||
XM_005260504.4 | XP_005260561.1 | 8, 9 | 2195 | |||
XM_011528979.2 | XP_011527281.1 | 7, 8 | 1850 | |||
XM_017028012.1 | XP_016883501.1 | 7, 8 | 1253 | |||
GenBank mRNA | AF395833.1 | - | 8, 9 | 1459 | ||
AK296989.1 | - | 5, 6 | 1053 | |||
BC041331.2 | - | 5, 6 | 697 | |||
BC142981.1 | - | 5, 6 | 697 |