Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022098.3 | NP_071381.1 | 2 | 225 | ||
GenBank mRNA | AK301635.1 | - | 2 | 224 | ||
AK310502.1 | - | 2 | 483 | |||
AK313770.1 | - | 2 | 186 | |||
AL365514.1 | - | 2 | 208 | |||
AL834310.1 | - | 2 | 175 | |||
BC001208.1 | - | 2 | 160 | 3502251 | ||
BC001681.2 | - | 2 | 192 | 2823656 | ||
BC004989.1 | - | 2 | 192 | 2823656 | ||
BP346902.1 | - | 2 | 229 | |||
BX648018.1 | - | 2 | 220 | |||
CR456442.1 | - | 2 | 161 | |||
CR457396.1 | - | 2 | 124 |