Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276501.1 | NP_001263430.1 | 3 | 722 | ||
NM_022107.2 | NP_071390.1 | 7 | 693 | |||
GenBank mRNA | AF155657.1 | - | 3 | 341 | ||
AJ243937.1 | - | 7 | 693 | |||
AK313922.1 | - | 3 | 722 | |||
BC018724.1 | - | 3 | 385 | 4576337 |