Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031231.3 | NP_112508.3 | 3 | 297 | ||
NM_031232.3 | NP_112509.3 | 3 | 297 | |||
XM_005260510.1 | XP_005260567.1 | 3 | 267 | |||
XM_011528992.2 | XP_011527294.1 | 3 | 267 | |||
GenBank mRNA | AF193759.1 | - | 3 | 204 | ||
AF409141.1 | - | 3 | 245 | |||
AK023706.1 | - | 3 | 248 | |||
AK291895.1 | - | 3 | 227 | |||
AK309285.1 | - | 3 | 259 | |||
AL519842.3 | - | 3 | 297 | |||
BC047673.1 | - | 3 | 252 | 5755231 | ||
BQ431858.1 | - | 3 | 243 |