Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031231.3 | NP_112508.3 | 5 | 445 | ||
NM_031232.3 | NP_112509.3 | 5 | 445 | |||
XM_005260510.1 | XP_005260567.1 | 5 | 415 | |||
XM_011528992.2 | XP_011527294.1 | 5 | 415 | |||
GenBank mRNA | AB039947.1 | - | 3 | 147 | ||
AF193759.1 | - | 5 | 352 | |||
AF409141.1 | - | 5 | 393 | |||
AK023706.1 | - | 5 | 396 | |||
AK291895.1 | - | 5 | 375 | |||
AK309285.1 | - | 5 | 829 | |||
AL519842.3 | - | 5 | 445 | |||
BC047673.1 | - | 5 | 400 | 5755231 | ||
BQ431858.1 | - | 5 | 391 |