Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040283.2 | NP_001035373.1 | 6, 7 | 849 | ||
XM_011527214.1 | XP_011525516.1 | 6, 7 | 861 | |||
XM_011527215.1 | XP_011525517.1 | 6, 7 | 853 | |||
XM_017027123.1 | XP_016882612.1 | 6, 7 | 866 | |||
XM_017027124.1 | XP_016882613.1 | 6, 7 | 868 | |||
XM_017027125.1 | XP_016882614.1 | 6, 7 | 871 | |||
XM_017027126.1 | XP_016882615.1 | 6, 7 | 961 | |||
XM_017027127.1 | XP_016882616.1 | 6, 7 | 826 | |||
XM_017027128.1 | XP_016882617.1 | 6, 7 | 1031 | |||
XM_017027129.1 | XP_016882618.1 | 6, 7 | 862 | |||
GenBank mRNA | AJ291669.1 | - | 5, 6 | 665 | ||
AK057404.1 | - | 6, 7 | 853 | |||
AK301822.1 | - | 6, 7 | 852 | |||
BC029202.1 | - | 6, 7 | 815 | 5170924 | ||
BC039266.1 | - | 6, 7 | 817 | 5298983 |