Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033067.2 | NP_149056.1 | 3 | 988 | ||
GenBank mRNA | AJ291671.1 | - | 3 | 915 | ||
AK057273.1 | - | 3 | 988 | |||
AK302085.1 | - | 4 | 527 | |||
BC029566.1 | - | 3 | 504 | 4836894 | ||
KU178739.1 | - | 3 | 474 | |||
KU178740.1 | - | 2 | 301 |