Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53AIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195194.1 | NP_001182123.1 | 2 | 237 | ||
NM_001195195.1 | NP_001182124.1 | 3 | 903 | |||
NM_001251964.1 | NP_001238893.1 | 2 | 1931 | |||
NM_022112.2 | NP_071395.2 | 3 | 903 | |||
XM_017018113.1 | XP_016873602.1 | 3 | 1668 | |||
XM_017018114.1 | XP_016873603.1 | 3 | 1674 | |||
XM_017018115.1 | XP_016873604.1 | 5 | 1847 | |||
XM_017018116.1 | XP_016873605.1 | 3 | 4933 | |||
XM_017018117.1 | XP_016873606.1 | 3 | 1673 | |||
GenBank mRNA | AB045830.1 | - | 3 | 431 | ||
AB045831.1 | - | 3 | 431 | |||
AB045832.1 | - | 2 | 1459 | |||
AK310612.1 | - | 3 | 1505 | |||
AY302135.1 | - | 2 | 221 |