Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024504.3 | NP_078780.1 | 2 | 741 | ||
GenBank mRNA | AF319458.1 | - | 2 | 741 | ||
AK022595.1 | - | 2 | 741 | |||
BC052311.1 | - | 2 | 654 | 6671984 | ||
KJ463837.1 | - | 1 | 539 |