Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313726.1 | NP_001300655.1 | 22, 23 | 2379 | ||
NM_001313727.1 | NP_001300656.1 | 18, 19 | 1937 | |||
NM_031418.3 | NP_113606.2 | 21, 22 | 2287 | |||
XM_011520282.2 | XP_011518584.1 | 8, 9 | 834 | |||
XM_017018118.1 | XP_016873607.1 | 19, 20 | 1915 | |||
XM_017018119.1 | XP_016873608.1 | 16, 17 | 1527 | |||
GenBank mRNA | AJ300461.1 | - | 21, 22 | 2994 | ||
AK295143.1 | - | 18, 19 | 1849 | |||
AK295658.1 | - | 5, 6 | 322 | |||
AK295816.1 | - | 18, 19 | 1849 | |||
AK302659.1 | - | 21, 22 | 2326 | |||
AK304781.1 | - | 22, 23 | 2220 | |||
AK316018.1 | - | 18, 19 | 1849 | |||
BQ430769.1 | - | 2, 3 | 158 |