Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033054.2 | NP_149043.2 | 20 | 2839 | ||
XM_017012503.1 | XP_016867992.1 | 19 | 2598 | |||
XM_017012504.1 | XP_016867993.1 | 20 | 2834 | |||
GenBank mRNA | AB290179.1 | - | 20 | 2799 | ||
AK074135.1 | - | 15 | 3003 | |||
AK131089.1 | - | 15 | 4956 | |||
AL832302.1 | - | 19 | 2594 | |||
BC015693.2 | - | 4 | 529 | |||
BC113544.1 | - | 20 | 2786 | 8322596 | ||
BC117241.1 | - | 20 | 2786 | 40125792 |