Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022122.2 | NP_071405.2 | 8 | 1119 | ||
XM_011542948.2 | XP_011541250.1 | 7 | 1038 | |||
XM_011542949.2 | XP_011541251.1 | 7 | 1005 | |||
XM_011542950.2 | XP_011541252.1 | 7 | 865 | |||
XM_017018120.1 | XP_016873609.1 | 8 | 1164 | |||
GenBank mRNA | AF195192.1 | - | 8 | 1102 | ||
AY358752.1 | - | 8 | 1119 | |||
BX102293.1 | - | 4 | 367 |