Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022126.3 | NP_071409.3 | 6 | 768 | ||
XM_005270026.3 | XP_005270083.1 | 6 | 717 | |||
XM_006717943.1 | XP_006718006.1 | 6 | 717 | |||
XM_017016509.1 | XP_016871998.1 | 6 | 717 | |||
XM_017016510.1 | XP_016871999.1 | 6 | 717 | |||
XM_017016511.1 | XP_016872000.1 | 6 | 717 | |||
XM_017016512.1 | XP_016872001.1 | 2 | 590 | |||
GenBank mRNA | AK055532.1 | - | 6 | 697 | ||
BC113629.1 | - | 6 | 768 | 8322681 | ||
BC113631.1 | - | 6 | 768 | 8322683 |