Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287580.1 | NP_001274509.1 | 8 | 1868 | ||
NM_022128.2 | NP_071411.1 | 7 | 749 | |||
GenBank mRNA | AJ404857.1 | - | 7 | 709 | ||
AK300989.1 | - | 7 | 703 | |||
AY643715.1 | - | 7 | 741 | |||
BC017425.1 | - | 7 | 751 | 4694872 |