Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033083.1 | NP_001028255.1 | 3 | 337 | ||
NM_022129.3 | NP_071412.2 | 3 | 337 | |||
XM_005270028.3 | XP_005270085.1 | 3 | 371 | |||
XM_011540060.2 | XP_011538362.1 | 3 | 371 | |||
XM_017016513.1 | XP_016872002.1 | 3 | 330 | |||
XM_017016514.1 | XP_016872003.1 | 3 | 371 | |||
GenBank mRNA | AB049758.1 | - | 1 | 199 | ||
AK027673.1 | - | 3 | 305 | |||
AK092826.1 | - | 2 | 169 | |||
AK312275.1 | - | 2 | 128 | |||
BC009738.1 | - | 3 | 319 | 3879159 | ||
CB121561.1 | - | 3 | 338 | |||
GQ472233.1 | - | 3 | 319 |