Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022130.3 | NP_071413.1 | 2, 3 | 636 | ||
XM_017009686.1 | XP_016865175.1 | 3, 4 | 905 | |||
XM_017009687.1 | XP_016865176.1 | 2, 3 | 730 | |||
GenBank mRNA | AJ296152.1 | - | 2, 3 | 457 | ||
AK057544.1 | - | 4, 5 | 585 | |||
AK075156.1 | - | 2, 3 | 636 | |||
AK092604.1 | - | 3, 4 | 497 | |||
AK293368.1 | - | 3, 4 | 425 | |||
AK301588.1 | - | 3, 4 | 543 | |||
AL133078.1 | - | 1, 2 | 83 | |||
BC012123.1 | - | 2, 3 | 591 | 4558305 | ||
BC033725.1 | - | 2, 3 | 628 | 4999629 | ||
BC063586.1 | - | 2, 3 | 560 | 4428707 |