Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022130.3 | NP_071413.1 | 2 | 579 | ||
XM_017009686.1 | XP_016865175.1 | 3 | 848 | |||
XM_017009687.1 | XP_016865176.1 | 2 | 673 | |||
GenBank mRNA | AJ296152.1 | - | 2 | 400 | ||
AK057544.1 | - | 4 | 528 | |||
AK075156.1 | - | 2 | 579 | |||
AK092604.1 | - | 3 | 440 | |||
AK293368.1 | - | 3 | 368 | |||
AK301588.1 | - | 3 | 486 | |||
AL133078.1 | - | 1 | 26 | |||
BC012123.1 | - | 2 | 534 | 4558305 | ||
BC033725.1 | - | 2 | 571 | 4999629 | ||
BC063586.1 | - | 2 | 503 | 4428707 |