Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022133.3 | NP_071416.2 | 8 | 1414 | ||
NM_152836.2 | NP_690049.1 | 7 | 1168 | |||
NM_152837.2 | NP_690050.1 | 6 | 1081 | |||
XM_005251282.4 | XP_005251339.1 | 8 | 1121 | |||
XM_005251283.2 | XP_005251340.1 | 9 | 1258 | |||
XM_011517574.2 | XP_011515876.1 | 6 | 511 | |||
GenBank mRNA | AF305779.1 | - | 7 | 1019 | ||
AK290903.1 | - | 7 | 1084 | |||
AL833763.1 | - | 6 | 988 | |||
BC033630.1 | - | 7 | 1076 | 5224216 |