Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMSN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256370.1 | NP_001243299.1 | 3 | 469 | ||
NM_001286523.1 | NP_001273452.1 | 3 | 313 | |||
NM_022136.4 | NP_071419.3 | 2 | 189 | |||
XM_011529684.2 | XP_011527986.1 | 3 | 229 | |||
XM_011529685.1 | XP_011527987.1 | 2 | 144 | |||
XM_011529686.1 | XP_011527988.1 | 2 | 186 | |||
XM_017028427.1 | XP_016883916.1 | 2 | 186 | |||
GenBank mRNA | AF218085.2 | - | 3 | 469 | ||
AF222927.1 | - | 2 | 172 | |||
AK125144.1 | - | 2 | 172 | |||
BC029112.1 | - | 2 | 163 | 4343284 | ||
DA819791.1 | - | 2 | 189 | |||
HG380513.1 | - | 3 | 219 | |||
HG380514.1 | - | 3 | 288 |