Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166412.1 | NP_001159884.1 | 4, 5 | 689 | ||
NM_022138.2 | NP_071421.1 | 4, 5 | 689 | |||
XM_011536065.1 | XP_011534367.1 | 4, 5 | 687 | |||
XM_011536066.1 | XP_011534368.1 | 4, 5 | 687 | |||
GenBank mRNA | AB014730.1 | - | 4, 5 | 489 | ||
AB014737.1 | - | 4, 5 | 489 | |||
AF173892.1 | - | 3, 4 | 344 | |||
AJ420521.1 | - | 4, 5 | 669 | |||
AK056700.1 | - | 4, 5 | 616 | |||
AK314613.1 | - | 4, 5 | 656 | |||
AL832303.1 | - | 4, 5 | 591 | |||
BC028420.1 | - | 4, 5 | 692 | 4826774 | ||
BC047583.1 | - | 4, 5 | 661 | 4826950 |