Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022165.2 | NP_071448.1 | 4 | 356 | ||
XM_006723323.3 | XP_006723386.1 | 3 | 304 | |||
GenBank mRNA | AF311862.1 | - | 4 | 334 | ||
AY358744.1 | - | 4 | 356 | |||
BC027618.2 | - | 4 | 330 | 4830017 |