Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022168.3 | NP_071451.2 | 12 | 2811 | ||
GenBank mRNA | AF095844.1 | - | 12 | 2578 | ||
AK056293.1 | - | 6 | 988 | |||
AK292941.1 | - | 13 | 2725 | |||
AK314611.1 | - | 12 | 2806 | |||
AY017378.1 | - | 12 | 2564 | |||
BC111750.1 | - | 12 | 2478 | 40008600 |