Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022168.3 | NP_071451.2 | 13 | 2919 | ||
GenBank mRNA | AF095844.1 | - | 13 | 2686 | ||
AK314611.1 | - | 13 | 2914 |