Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1AIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022347.3 | NP_071742.1 | 3 | 977 | ||
XM_017000477.1 | XP_016855966.1 | 2 | 1650 | |||
XM_017000478.1 | XP_016855967.1 | 2 | 983 | |||
GenBank mRNA | AJ299406.1 | - | 1 | 50 | ||
AK289793.1 | - | 3 | 952 | |||
BC032790.1 | - | 1 | 251 | |||
BC096704.1 | - | 1 | 50 | 40021613 | ||
BC098170.1 | - | 1 | 50 | 40021614 | ||
BC098309.1 | - | 1 | 50 | 40021617 | ||
BC098348.1 | - | 1 | 50 | 40021612 |