Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022353.2 | NP_071748.2 | 6, 7 | 1621 | ||
XM_005246766.3 | XP_005246823.1 | 6, 7 | 1221 | |||
XM_006712685.1 | XP_006712748.1 | 6, 7 | 1333 | |||
XM_006712686.1 | XP_006712749.1 | 6, 7 | 1204 | |||
XM_011511631.1 | XP_011509933.1 | 6, 7 | 1352 | |||
XM_017004676.1 | XP_016860165.1 | 6, 7 | 1221 | |||
XM_017004677.1 | XP_016860166.1 | 6, 7 | 1221 | |||
XM_017004678.1 | XP_016860167.1 | 6, 7 | 1333 | |||
XM_017004679.1 | XP_016860168.1 | 6, 7 | 1352 | |||
XM_017004680.1 | XP_016860169.1 | 6, 7 | 1307 | |||
XM_017004681.1 | XP_016860170.1 | 6, 7 | 1176 | |||
XM_017004682.1 | XP_016860171.1 | 6, 7 | 1204 | |||
XM_017004683.1 | XP_016860172.1 | 6, 7 | 1204 | |||
XM_017004684.1 | XP_016860173.1 | 6, 7 | 1113 | |||
XM_017004685.1 | XP_016860174.1 | 6, 7 | 1342 | |||
GenBank mRNA | AJ295148.1 | - | 4, 5 | 1185 | ||
AK314565.1 | - | 6, 7 | 1621 | |||
BC011904.2 | - | 6, 7 | 1190 | 4121450 |